missing translation for 'onlineSavingsMsg'
Learn More

HSP10/EPF Rabbit anti-Human, Mouse, Rat, Clone: 0H0X6, Novus Biologicals™

Product Code. 18331113 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331113 20 μg 20µL
18369703 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18331113 Supplier Novus Biologicals Supplier No. NBP31660420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HSP10/EPF Monoclonal antibody specifically detects HSP10/EPF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP10/EPF
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 0H0X6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 10 kDa chaperonin, 10 kDa heat shock protein, mitochondrial, Chaperonin 10, CPN10HSP10, Early-pregnancy factor, EPF, GROES, heat shock 10kD protein 1 (chaperonin 10), heat shock 10kDa protein 1 (chaperonin 10), Hsp10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-150 of human HSP10/EPF (P61604).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Membrane Trafficking and Chaperones, Ovarian Carcinoma Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3336
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.