missing translation for 'onlineSavingsMsg'
Learn More

HSP10/EPF Antibody (330629) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30731596 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30731596 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731596 Supplier Novus Biologicals Supplier No. FAB3298I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HSP10/EPF Monoclonal antibody specifically detects HSP10/EPF in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP10/EPF
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 330629
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias 10 kDa chaperonin, 10 kDa heat shock protein, mitochondrial, Chaperonin 10, CPN10HSP10, Early-pregnancy factor, EPF, GROES, heat shock 10kD protein 1 (chaperonin 10), heat shock 10kDa protein 1 (chaperonin 10), Hsp10
Host Species Mouse
Immunogen E. coli-derived recombinant human HSP10/EPF, Ala2-Asp102, Accession # P61604
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Membrane Trafficking and Chaperones, Ovarian Carcinoma Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3336
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.