Learn More
Invitrogen™ HSL Polyclonal Antibody
Chicken Polyclonal Antibody
Brand: Invitrogen™ PA19580
Description
PA1-9580 detects HSL in mouse samples. PA1-9580 has been successfully used in Western blot, immunohistochemistry, and immunocytochemistry procedures. By Western blot, PA1-9580 detects a ~83 kDa band representing HSL, with a non-specific band at ~40 kDa. PA1-9580 immunogen is (LSGRKPQKTTSPTAES) mouse HSL protein.
Hormone-sensitive lipase (HSL) is an intracellular neutral lipase found in a variety of tissues, but primarily in adipose and steroidogenic tissues. It hydrolyzes triglycerides and cholesteryl esters. It is the major enzyme responsible for the mobilization of free fatty acids from adopise tissue.
Specifications
| HSL | |
| Polyclonal | |
| Unconjugated | |
| LIPE | |
| 4933403G17Rik; AOMS4; EC 3.1.1.79; FPLD6; hormone - sensitive lipase testicular isoform; hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; HSL antibody; LHS; lipase E; lipase E, hormone sensitive type; lipase, hormone sensitive; lipase, hormone-sensitive; LIPE; LIPS | |
| Chicken | |
| Affinity Chromatography | |
| RUO | |
| 16890 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.02% sodium azide | |
| P54310 | |
| LIPE | |
| Mouse HSL peptide (LSGRKPQKTTSPTAES). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgY |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.