missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSFY1/HSFY2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528352
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: mouse testis, Rat testis. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the heat shock factor (HSF) family of transcriptional activators for heat shock proteins. This gene is a candidate gene for azoospermia, since it localizes to a region of chromosome Y that is sometimes deleted in infertile males. The genome has two identical copies of this gene within a palindromic region; this record represents the more centromeric copy. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
HSFY1/HSFY2 | |
Polyclonal | |
Unconjugated | |
HSFY1 | |
4933413G11Rik; Heat shock transcription factor 2-like protein; heat shock transcription factor, Y linked 2; heat shock transcription factor, Y linked 2 like; heat shock transcription factor, Y-linked; heat shock transcription factor, Y-linked 1; heat shock transcription factor, Y-linked 2; HSF2L; HSF2-like; HSFY; Hsfy1; HSFY2; Hspy2l | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
159119, 316409, 71066, 86614 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96LI6 | |
HSFY1, Hsfy2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 401 of Human HSFY1+2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |