Learn More
Invitrogen™ HSFX1 Monoclonal Antibody (1D7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521924
Description
Peptide Sequence: MEDKRSLSMA RCEERNSRGQ DHGLERVPFP PQLQSETYLH PADPSPAWDD PGSTGSPNLR LLTEEIAFQP LAEEASFRRP HPDGDVPPQG EDNLLSLPFP QKLWRLVSSN QFSSIWWDDS GACRVINQKL FEKEILKRDV AHKVFATTSI KSFFRQLNLY GFRKRRQCTF RTFTRIFSAK RLVSILNKLE FYCHPYFQRD SPHLLVRMKR RVGVKSAPRH QEEDKPEAAG SCLAPADTEQ QDHTSPNEND QVTPQHREPA GPNTQIRSGS APPATPVMVP DSAVASDNSP VTQPAGEWSE GSQAHVTPVA AVPGPAALPF LYVPGSPTQM NSYGPVVALP TASRSTLAMD TTGLPAPGML PFCHLWVPVT LVAAGAAQPA ASMVMFPHLP ALHHHCPHSH RTSQYMPASD GPQAYPDYAD QST.
HSFX1 (Heat Shock Transcription Factor Family, X-Linked 1) is a Protein Coding gene. Diseases associated with HSFX1 include Microsporidiosis. Gene Ontology (GO) annotations related to this gene include DNA-binding transcription factor activity and sequence-specific DNA binding. An important paralog of this gene is HSFX2. [GeneCards].
Specifications
| HSFX1 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9UBD0 | |
| HSFX1 | |
| HSFX1 (AAH21706, 1 a.a. ~ 423 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1D7 | |
| Unconjugated | |
| HSFX1 | |
| heat shock transcription factor family, X linked 1; heat shock transcription factor family, X-linked 1; heat shock transcription factor, X-linked; HSFX1; HSFX2; LW-1; ORC4L; testicular secretory protein Li 21 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 100506164 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.