missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564126
This item is not returnable.
View return policy
Description
Immunogen sequence: LNHNPSPSSV VFVQEGPPFS THQVDANIKC QTSSRENILP SEQMGFLISE MGPASKPSED TGLATPARYR EHRSNSQQGK SPDLH Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 80%.
HSF5 may act as a transcriptional factor.
Specifications
| HSF5 | |
| Polyclonal | |
| Unconjugated | |
| Hsf5 | |
| Gm739; heat shock factor protein 5; Heat shock transcription factor 5; heat shock transcription factor family member 5; HSF 5; HSF5; HSTF 5; HSTF5; LOW QUALITY PROTEIN: heat shock factor protein 5; MLZ-220; Protein expressed in male leptotene and zygotene spermatocytes 220 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 124535 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4G112 | |
| Hsf5 | |
| Recombinant protein corresponding to Human HSF5. Recombinant protein control fragment (Product #RP-103965). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction