Learn More
HSF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22883-1-AP-20UL
 View more versions of this product
Product Code. 16862125
  • £135.00 / 20µL
Description
This antibody is specific to human HSF4.
Heat-shock transcription factors (HSFs) activate heat-shock response genes under conditions of heat or other stresses. HSF4 lacks the carboxyl-terminal hydrophobic repeat which is shared among all vertebrate HSFs and has been suggested to be involved in the negative regulation of DNA binding activity. Two alternatively spliced transcripts encoding distinct isoforms and possessing different transcriptional activity have been described.
Specifications
Specifications
HSF4
Polyclonal
Unconjugated
HSF4
CTM, Heat shock factor protein 4, hHSF4, HSF 4, HSF4, HSTF 4
Rabbit
Antigen Affinity Chromatography
RUO
26386, 291960, 3299
-20°C
Liquid
Western Blot
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9R0L1, Q9ULV5
Hsf4
HSF4 Fusion Protein Ag18935
20 μL
Primary
Mouse, Rat, Human