missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18797-1-AP-150UL
This item is not returnable.
View return policy
Description
Heat-shock transcription factors (HSFs) activate heat-shock response genes under conditions of heat or other stresses. HSF4 lacks the carboxyl-terminal hydrophobic repeat which is shared among all vertebrate HSFs and has been suggested to be involved in the negative regulation of DNA binding activity. Two alternatively spliced transcripts encoding distinct isoforms and possessing different transcriptional activity have been described.Specifications
| HSF4 | |
| Polyclonal | |
| Unconjugated | |
| HSF4 | |
| CTM, Heat shock factor protein 4, hHSF4, HSF 4, HSF4, HSTF 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26386, 3299 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9R0L1, Q9ULV5 | |
| HSF4 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction