missing translation for 'onlineSavingsMsg'
Learn More

HSF4 Antibody (2A2), Novus Biologicals™

Product Code. 18388317 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388317 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18388317 Supplier Novus Biologicals Supplier No. H00003299M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HSF4 Monoclonal antibody specifically detects HSF4 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSF4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2A2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001529
Gene Alias cataract, Marner, heat shock factor protein 4, heat shock transcription factor 4CTM, hHSF4, HSF 4, HSTF 4
Host Species Mouse
Immunogen HSF4 (NP_001529, 121 a.a. ~ 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KVPALRGDDGRWRPEDLGRLLGEVQALRGVQESTEARLRELRQQNEILWREVVTLRQSHGQQHRVIGKLIQCLFGPLQAGPSNAGGKRKLSLMLDEGSSC
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3299
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.