Learn More
Invitrogen™ HSF2BP Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA540978
Product Code. 15564532
  • £441.00 / 100µL
Description
Peptide sequence: RHMGTKEEFV KVRKKDLERL TTEVMQIRDF LPRILNGEVL ESFQKLKIVE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.
HSF2 binding protein (HSF2BP) associates with HSF2. The interaction occurs between the trimerization domain of HSF2 and the amino terminal hydrophilic region of HSF2BP that comprises two leucine zipper motifs. HSF2BP may therefore be involved in modulating HSF2 activation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HSF2BP
Polyclonal
Unconjugated
HSF2BP
4932437G14Rik; heat shock factor 2-binding protein; heat shock transcription factor 2 binding protein; Hsf2bp; RGD1565879
Rabbit
Protein A
RUO
11077
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
O75031
HSF2BP
Synthetic peptide directed towards the N-terminal of human HSF2BP (aa 12-61).