missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549820
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total HSF2 protein.
Prokaryotic and eukaryotic cells respond to thermal and chemical stress by inducing a group of genes collectively designated heat shock genes. In eukaryotes, this gene expression is regulated primarily at the transcription level. Heat shock transcription factors (HSF, also designated HSTF) 1 and 2 are involved in this regulation. HSF1 and HSF2 are upregulated by estrogen at both the mRNA and protein level. HSF1 is normally found as a monomer whose transcriptional activity is repressed by constitutive phosphorylation. Upon activation, HSF1 forms trimers, gains DNA binding activity and is translocated to the nucleus. HSF2 activity is associated with differentiation and development, and, like HSF1, binds DNA as a trimer. Both HSF1 and HSF2 are known to be induced by proteasome inhibitors of the ubiquitin pathway.
Specifications
| HSF2 | |
| Polyclonal | |
| Unconjugated | |
| HSF2 | |
| AI661205; heat shock factor 2; heat shock factor protein 2; Heat shock transcription factor 2; HSF 2; Hsf2; HSTF 2; HSTF2; MGC117376; MGC156196; MGC75048 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15500, 3298, 64441 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P38533, Q03933 | |
| HSF2 | |
| Synthesized peptide derived from internal of human HSF2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction