missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSF2 Monoclonal Antibody (OTI2A7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526871
This item is not returnable.
View return policy
Description
HSF2 Monoclonal Antibody for Western Blot, IHC (P)
Prokaryotic and eukaryotic cells respond to thermal and chemical stress by inducing a group of genes collectively designated heat shock genes. In eukaryotes, this gene expression is regulated primarily at the transcription level. Heat shock transcription factors (HSF, also designated HSTF) 1 and 2 are involved in this regulation. HSF1 and HSF2 are upregulated by estrogen at both the mRNA and protein level. HSF1 is normally found as a monomer whose transcriptional activity is repressed by constitutive phosphorylation. Upon activation, HSF1 forms trimers, gains DNA binding activity and is translocated to the nucleus. HSF2 activity is associated with differentiation and development, and, like HSF1, binds DNA as a trimer. Both HSF1 and HSF2 are known to be induced by proteasome inhibitors of the ubiquitin pathway.
Specifications
| HSF2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q03933 | |
| HSF2 | |
| Human recombinant protein fragment corresponding to amino acids 74-324 of HSF2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A7 | |
| Unconjugated | |
| HSF2 | |
| AI661205; heat shock factor 2; heat shock factor protein 2; Heat shock transcription factor 2; HSF 2; Hsf2; HSTF 2; HSTF2; MGC117376; MGC156196; MGC75048 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3298 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction