missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF2 Antibody (SR2042), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | HSF2 |
|---|---|
| Clone | SR2042 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18676504
|
Novus Biologicals
NBP3-22062-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600874
|
Novus Biologicals
NBP3-22062-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSF2 Monoclonal antibody specifically detects HSF2 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| HSF2 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| heat shock factor 2, heat shock factor protein 2, heat shock transcription factor 2MGC75048, HSF 2, HSTF 2, HSTF2, MGC117376, MGC156196 | |
| A synthesized peptide derived from human HSF2 (Uniprot #: Q03933) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2042 | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cellular Signaling, DNA replication Transcription Translation and Splicing, Epigenetics | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 3298 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title