missing translation for 'onlineSavingsMsg'
Learn More

HSF1A, MedChemExpress

Product Code. 16476487
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476487

Brand: Medchem Express HY103000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HSF1A is a cell-permeable activator of heat shock transcription factor 1 (HSF1). HSF1A also acts as a specific inhibitor of TRiC/CCT. Chaperonin TCP-1 ring complex (TRiC)/chaperonin containing TCP-1 (CCT) plays a pivotal role in toxin translocation and/or refolding.

HSF1A is a cell-permeable activator of heat shock transcription factor 1 (HSF1). HSF1A also acts as a specific inhibitor of TRiC/CCT. Chaperonin TCP-1 ring complex (TRiC)/chaperonin containing TCP-1 (CCT) plays a pivotal role in toxin translocation and/or refolding.

Specifications

Chemical Name or Material HSF1A
CAS 1196723-93-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H19N3O2S2
Quantity 5 mg
Solubility Information DMSO : ≥ 150 mg/mL (366.28 mM)
SMILES O=S(C1=CC=C(CC)C=C1)(NC2=CC(C3=CC=CS3)=NN2C4=CC=CC=C4)=O
Molecular Weight (g/mol) 409.52
Formula Weight 409.52
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.