missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16107-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5′ UTR, but encoding the same protein, have been found for this gene.Specifications
| HSF1 | |
| Polyclonal | |
| Unconjugated | |
| HSF1 | |
| Heat shock factor protein 1, HSF 1, HSF1, HSTF 1, HSTF1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15499, 3297, 79245 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P38532, Q00613 | |
| HSF1 | |
| HSF1 Fusion Protein Ag9023 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction