missing translation for 'onlineSavingsMsg'
Learn More

HSF1 Rabbit anti-Human, Clone: S07-0G0, Novus Biologicals™

Product Code. 18312085 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18312085 25 μg 25µL
18372392 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18312085 Supplier Novus Biologicals Supplier No. NBP31489525UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HSF1 Monoclonal antibody specifically detects HSF1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Gel Supershift Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSF1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Gel Shift
Classification Monoclonal
Clone S07-0G0
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100 - 1:2000, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Supershift Assay
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Host Species Rabbit
Immunogen A synthetic peptide of human HSF1 (Uniprot#: Q00613)
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3297
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.