missing translation for 'onlineSavingsMsg'
Learn More

HSD5 Antibody, Novus Biologicals™

Product Code. 18395262 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395262 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395262 Supplier Novus Biologicals Supplier No. NBP312203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HSD5 Polyclonal antibody specifically detects HSD5 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD5
Applications Western Blot, ELISA, Immunoprecipitation, Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:2000, Immunoprecipitation 1:200, Dot Blot 1:2000
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias chromosome 3 open reading frame 34, hypothetical protein LOC84984, MGC14126
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 120-170 on human Chromosome open reading frame 34 protein.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84984
Target Species Human
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.