missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSD3B7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562204
This item is not returnable.
View return policy
Description
Immunogen sequence: VVRMLLQREP RLGELRVFDQ HLGPWLEELK TGPVRVTAIQ GDVTQAHEVA AAVAGAHVVI HTAGLVDVFG RASPKTIHEV NVQG Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
This gene encodes an enzyme which is involved in the initial stages of the synthesis of bile acids from cholesterol and a member of the short-chain dehydrogenase/reductase superfamily. The encoded protein is a membrane-associated endoplasmic reticulum protein which is active against 7-alpha hydrosylated sterol substrates. Mutations in this gene are associated with a congenital bile acid synthesis defect which leads to neonatal cholestasis, a form of progressive liver disease. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Dec 2008].
Specifications
| HSD3B7 | |
| Polyclonal | |
| Unconjugated | |
| HSD3B7 | |
| 3 beta-hydroxy-delta 5-C27-steroid oxidoreductase; 3 beta-hydroxysteroid dehydrogenase type 7; 3 beta-hydroxysteroid dehydrogenase type VII; 3-beta-HSD VII; 3-beta-hydroxy-Delta(5)-C27 steroid oxidoreductase; AI195443; BB098564; C(27) 3-beta-HSD; C(27)-3BETA-HSD; CBAS1; Cca2; cholest-5-ene-3-be; cholest-5-ene-3-beta,7-alpha-diol 3-beta-dehydrogenase; Confluent 3Y1 cell-associated 2; HSD3B7; hsd3b7 {ECO:0000312; hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7; PFIC4; RGD:628727}; SDR11E3; short chain dehydrogenase/reductase family 11E, member 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2F3 | |
| HSD3B7 | |
| Recombinant protein corresponding to Human HSD3B7. Recombinant protein control fragment (Product #RP-101137). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction