missing translation for 'onlineSavingsMsg'
Learn More

HSD3B1 Antibody, Novus Biologicals™

Product Code. 18418991 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418991 25 μL 25µL
18184714 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18418991 Supplier Novus Biologicals Supplier No. NBP23338525ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HSD3B1 Polyclonal specifically detects HSD3B1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD3B1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P14060
Gene Alias 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I, 3BETAHSD, 3-beta-HSD I, 3-beta-hydroxy-Delta(5)-steroid dehydrogenase, 3BH, delta-5-3-ketosteroid isomerase, EC 1.1.1, HSD3B, HSDB3, HSDB3A, hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1, hydroxy-delta-5-steroid dehydrogenase, 3 beta-and steroid delta-isomerase 1, I, progesterone reductase, SDR11E1, short chain dehydrogenase/reductase family 11E, member 1,3-beta-hydroxy-5-ene steroid dehydrogenase, steroid Delta-isomerase, Trophoblast antigen FDO161G
Gene Symbols HSD3B1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: RGQFYYISDDTPHQSYDNLNYTLSKEFGLRLDSRWS
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Prostate Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3283
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.