missing translation for 'onlineSavingsMsg'
Learn More

HSD3B1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18358843 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358843 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358843 Supplier Novus Biologicals Supplier No. NBP299334JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HSD3B1 Polyclonal specifically detects HSD3B1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD3B1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I, 3BETAHSD, 3-beta-HSD I, 3-beta-hydroxy-Delta(5)-steroid dehydrogenase, 3BH, delta-5-3-ketosteroid isomerase, EC 1.1.1, HSD3B, HSDB3, HSDB3A, hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1, hydroxy-delta-5-steroid dehydrogenase, 3 beta-and steroid delta-isomerase 1, I, progesterone reductase, SDR11E1, short chain dehydrogenase/reductase family 11E, member 1,3-beta-hydroxy-5-ene steroid dehydrogenase, steroid Delta-isomerase, Trophoblast antigen FDO161G
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human HSD3B1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Prostate Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3283
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.