missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD3B1 Antibody (FDO66Q), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-78644B
This item is not returnable.
View return policy
Description
HSD3B1 Monoclonal specifically detects HSD3B1 in Human, Rat, Porcine, Bovine, Goat, Primate, Baboon samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| HSD3B1 | |
| Monoclonal | |
| Biotin | |
| 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I, 3BETAHSD, 3-beta-HSD I, 3-beta-hydroxy-Delta(5)-steroid dehydrogenase, 3BH, delta-5-3-ketosteroid isomerase, EC 1.1.1, HSD3B, HSDB3, HSDB3A, hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1, hydroxy-delta-5-steroid dehydrogenase, 3 beta-and steroid delta-isomerase 1, I, progesterone reductase, SDR11E1, short chain dehydrogenase/reductase family 11E, member 1,3-beta-hydroxy-5-ene steroid dehydrogenase, steroid Delta-isomerase, Trophoblast antigen FDO161G | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, baboon, bovine, rat and marmoset. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| FDO66Q | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| HSD3B1 | |
| JEG Choriocarcinoma cells | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Prostate Cancer, Signal Transduction | |
| 3283 | |
| Human, Rat, Pig, Bovine, Goat, Primate, Baboon | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction