missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD3B1 Antibody (FDO66Q) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80781
This item is not returnable.
View return policy
Description
HSD3B1 Monoclonal specifically detects HSD3B1 in Human, Rat, Porcine, Bovine, Goat, Primate, Baboon samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| HSD3B1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry : 10-20 μg/mL, Immunocytochemistry/Immunofluorescence : 10-20 μg/mL, Immunohistochemistry-Paraffin : 10-20 μg/mL, Immunohistochemistry-Frozen : 10-20 μg/mL | |
| 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I, 3BETAHSD, 3-beta-HSD I, 3-beta-hydroxy-Delta(5)-steroid dehydrogenase, 3BH, delta-5-3-ketosteroid isomerase, EC 1.1.1, HSD3B, HSDB3, HSDB3A, hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1, hydroxy-delta-5-steroid dehydrogenase, 3 beta-and steroid delta-isomerase 1, I, progesterone reductase, SDR11E1, short chain dehydrogenase/reductase family 11E, member 1,3-beta-hydroxy-5-ene steroid dehydrogenase, steroid Delta-isomerase, Trophoblast antigen FDO161G | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, baboon, bovine, rat and marmoset. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| FDO66Q | |
| Unconjugated | |
| Tris-Glycine, 0.15 M NaCl | |
| HSD3B1 | |
| JEG Choriocarcinoma cells | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Prostate Cancer, Signal Transduction | |
| 3283 | |
| Human, Rat, Pig, Bovine, Goat, Primate, Baboon | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction