missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSD17B8 Monoclonal Antibody (OTI6H2)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526466
This item is not returnable.
View return policy
Description
HSD17B8 Monoclonal Antibody for Western Blot, IHC (P)
NAD-dependent 17-beta-hydroxysteroid dehydrogenase with highest activity towards estradiol. Has very low activity towards testosterone. The heteroteramer with CBR4 has NADH-dependent 3- ketoacyl-acyl carrier protein reductase activity. May play a role in biosynthesis of fatty acids in mitochondria.
Specifications
| HSD17B8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5TJF5, Q92506 | |
| HSD17B8 | |
| Human recombinant protein fragment corresponding to amino acids 57-261 of HSD17B8 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Canine | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6H2 | |
| Unconjugated | |
| HSD17B8 | |
| 17-beta-HSD 8; 17-beta-hydroxysteroid dehydrogenase 8; 3-ketoacyl-[acyl-carrier-protein] reductase alpha subunit; 3-oxoacyl-[acyl-carrier-protein] reductase; D17H6S112E; D6S2245E; dJ1033B10.9; Estradiol 17-beta-dehydrogenase 8; estrogen 17-oxidoreductase; FABG; FABGL; H FABGL; H2-K region expressed gene 6; H-2Ke6; H2-KE6; Hke6; Hsd17b8; hydroxysteroid (17-beta) dehydrogenase 8; hydroxysteroid 17-beta dehydrogenase 8; KAR alpha subunit; KE6; Ke-6; Protein Ke6; really interesting new gene 2 protein; RING2; SDR30C1; Short chain dehydrogenase/reductase family 30C member 1; short chain dehydrogenase/reductase family 30C, member 1; Testosterone 17-beta-dehydrogenase 8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 607895, 7923 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction