missing translation for 'onlineSavingsMsg'
Learn More

HSD17B8 Antibody, Novus Biologicals™

Product Code. 18390229 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390229 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390229 Supplier Novus Biologicals Supplier No. H00007923B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

HSD17B8 Polyclonal antibody specifically detects HSD17B8 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH08185
Gene Alias D6S2245EdJ1033B10.9, EC 1.1.1.-, EC 1.1.1.62, EC 1.1.1.63,17-beta-HSD 8, estrogen 17-oxidoreductase, FABGLestradiol 17-beta-dehydrogenase 8, H2-KE6, HKE6FabG (beta-ketoacyl-[acyl-carrier-protein] reductase, E coli) like (E. coli), hydroxysteroid (17-beta) dehydrogenase 8,3-oxoacyl-[acyl-carrier-protein] reductase, ke-6, KE6, Ke-6,17-beta-hydroxysteroid dehydrogenase 8, Protein Ke6, Really interesting new gene 2 protein, RING2FABG, SDR30C1, short chain dehydrogenase/reductase family 30C, member 1, Testosterone 17-beta-dehydrogenase 8
Host Species Mouse
Immunogen HSD17B8 (AAH08185, 1 a.a. - 261 a.a.) full-length human protein. MASQLQNRLRSALALVTGAGSGIGRAVSVRLAGEGATVAACDLDRAAAQETVRLLGGPGSKEGPPRGNHAAFQADVSEARAARCLLEQVQACFSRPPSVVVSCAGITQDEFLLHMSEDDWDKVIAVNLKGTFLVTQAAAQALVSNGCRGSIINISSIVGKVGNVGQINYAASKAGVIGLTQTAARELGRHGIRCNSVLPGFIATPMTQKVPQKVVDKITEMIPMGHLGDPEDVADVVAFLASEDSGYITGTSVEVTGGLFM
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7923
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.