missing translation for 'onlineSavingsMsg'
Learn More

HSD17B7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16823724 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16823724 20 μL 20µL
16813724 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16823724 Supplier Proteintech Supplier No. 148541AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Responsible for the reduction of the keto group on the C-3 of sterols.

Specifications

Antigen HSD17B7
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene HSD17B7
Gene Accession No. O88736, P56937, Q62904
Gene Alias 17 beta HSD 7, 3 keto steroid reductase, HSD17B7, PRAP, SDR37C1
Gene Symbols Hsd17b7
Host Species Rabbit
Immunogen HSD17B7 Fusion Protein Ag6639
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15490, 29540, 51478
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.