missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14854-1-AP-150UL
This item is not returnable.
View return policy
Description
Responsible for the reduction of the keto group on the C-3 of sterols.Specifications
| HSD17B7 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B7 | |
| 17 beta HSD 7, 3 keto steroid reductase, HSD17B7, PRAP, SDR37C1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15490, 29540, 51478 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O88736, P56937, Q62904 | |
| Hsd17b7 | |
| HSD17B7 Fusion Protein Ag6639 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction