missing translation for 'onlineSavingsMsg'
Learn More

HSD17B7 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18386956 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386956 100 μg 100µL
18325525 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18386956 Supplier Novus Biologicals Supplier No. NBP315527100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HSD17B7 Polyclonal antibody specifically detects HSD17B7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B7
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 50% glycerol, pH7.3
Gene Alias 17 beta-hydroxysteroid dehydrogenase type VII, 17-beta-hydroxysteroid dehydrogenase 7, 3-keto-steroid reductase, EC 1.1.1.270, EC 1.1.1.62, Estradiol 17-beta-dehydrogenase 7, hydroxysteroid (17-beta) dehydrogenase 7,17beta hydroxysteroid dehydrogenase, MGC12523, MGC75018,17-beta-HSD 7, PRAP, SDR37C1, short chain dehydrogenase/reductase family 37C, member 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 110-200 of human HSD17B7 (NP_057455.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51478
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.