missing translation for 'onlineSavingsMsg'
Learn More

HSD17B6 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18354713 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354713 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354713 Supplier Novus Biologicals Supplier No. NBP299211JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HSD17B6 Polyclonal specifically detects HSD17B6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B6
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 17-beta-HSD 6, 17-beta-HSD6, 17-beta-hydroxysteroid dehydrogenase type 6, 3(alpha->beta)-hydroxysteroid epimerase, 3-alpha->beta-HSE, EC 1.1.1, EC 1.1.1.105, EC 1.1.1.62, EC 1.1.1.63, HSE3-hydroxysteroid epimerase, hydroxysteroid (17-beta) dehydrogenase 6, hydroxysteroid (17-beta) dehydrogenase 6 homolog (mouse), NAD+ -dependent 3 alpha-hydroxysteroid dehydrogenase 3-hydroxysteroid epimerase, Oxidative 3-alpha hydroxysteroid dehydrogenase, oxidative 3-alpha-hydroxysteroid-dehydrogenase, oxidoreductase, retinol dehydrogenase, RODH3(alpha->beta)-hydroxysteroid epimerasel, SDR9C6, short chain dehydrogenase/reductase family 9C, member 6,3-alpha->beta-hydroxysteroid epimerase
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human HSD17B6.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8630
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.