missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15116-1-AP-150UL
This item is not returnable.
View return policy
Description
Peroxisomal multifunctional enzyme type 2 is a protein that in humans is encoded by the HSD17B4 gene. The protein encoded by this gene is a bifunctional enzyme that is involved in the peroxisomal beta-oxidation pathway for fatty acids. It also acts as a catalyst for the formation of 3-ketoacyl-CoA intermediates from both straight-chain and 2-methyl-branched-chain fatty acids. Defects in this gene that affect the peroxisomal fatty acid beta-oxidation activity are a cause of D-bifunctional protein deficiency (DBPD). An apparent pseudogene of this gene is present on chromosome 8. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
| HSD17B4 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B4 | |
| 17 beta HSD 4, D bifunctional protein, DBP, EDH17B4, HSD17B4, MFE 2, SDR8C1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15488, 3295, 79244 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51659, P51660, P97852 | |
| Hsd17b4 | |
| HSD17B4 Fusion Protein Ag7165 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction