missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSD17B4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579404
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human RT4 whole cell, rat liver tissue, mouse liver tissue. IHC: human intestinal cancer tissue, human lung cancer tissue, human mammary cancer tissue.
Peroxisomal multifunctional enzyme type 2 is a protein that in humans is encoded by the HSD17B4 gene. The protein encoded by this gene is a bifunctional enzyme that is involved in the peroxisomal beta-oxidation pathway for fatty acids. It also acts as a catalyst for the formation of 3-ketoacyl-CoA intermediates from both straight-chain and 2-methyl-branched-chain fatty acids. Defects in this gene that affect the peroxisomal fatty acid beta-oxidation activity are a cause of D-bifunctional protein deficiency (DBPD). An apparent pseudogene of this gene is present on chromosome 8. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| HSD17B4 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B4 | |
| (3R)-hydroxyacyl-CoA dehydrogenase; 17[b]-HSD; 17beta-estradiol dehydrogenase type IV; 17-beta-HSD; 17-beta-HSD 4; 17-beta-HSD IV; 17-beta-hydroxysteroid dehydrogenase 4; 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase; 4; beta-hydroxyacyl dehydrogenase; beta-keto-reductase; D-3-hydroxyacyl-CoA dehydratase; D-bifunctional protein; D-bifunctional protein, peroxisomal; DBP; Edh17b4; Enoyl-CoA hydratase 2; Hsd17b4; hydroxysteroid (17-beta) dehydrogenase 4; hydroxysteroid 17-beta dehydrogenase 4; hydroxysteroid dehydrogenase 4; MFE-2; MFP2; Mfp-2; MPF-2; Multifunctional protein 2; perMFE-2; peroxisomal multifunctional enzyme type 2; peroxisomal multifunctional enzyme type II; peroxisomal multifunctional protein 2; PRLTS1; SDR8C1; Short chain dehydrogenase/reductase family 8C member 1; short chain dehydrogenase/reductase family 8C, member 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15488, 3295, 79244 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P51659, P51660, P97852 | |
| HSD17B4 | |
| E.coli-derived human HSD17B4 recombinant protein (Position: D510-L736). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction