missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13415-1-AP-20UL
This item is not returnable.
View return policy
Description
This isoform of 17 beta-hydroxysteroid dehydrogenase is expressed predomitly in the testis and catalyzes the conversion of androstenedione to testosterone. It preferentially uses NADP as cofactor. Deficiency can result in male pseudohermaphroditism with gynecomastia.Specifications
| HSD17B3 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B3 | |
| 17 beta HSD 3, EDH17B3, HSD17B3, SDR12C2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117182, 15487, 3293 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O54939, P37058, P70385 | |
| HSD17B3 | |
| HSD17B3 Fusion Protein Ag4081 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction