missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSD17B2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554523
This item is not returnable.
View return policy
Description
Immunogen sequence: AVTMFSSVMR LELSKWGIKV ASIQPGGFLT NIAGTSDKWE KLEKDILDHL PAEVQEDYGQ DYILAQRNFL LLINSLASKD FSPVL Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 52%.
HSD17B2 is capable of catalyzing the interconversion of testosterone and androstenedione, as well as estradiol and estrone. HSD17B2 also has 20-alpha-HSD activity. HSD17B2 uses NADH while EDH17B3 uses NADPH.
Specifications
| HSD17B2 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B2 | |
| 1.1.1.239; 17 beta HSD 2; 17 HSD type 2; 17-beta hydroxysteroid dehydrogenase type 2; 17-beta-HSD 2; 17-beta-hydroxysteroid dehydrogenase 2; 17-beta-hydroxysteroid dehydrogenase type 2; 20 alpha HSD; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; AI194836; AI194967; AI255511; E2DH; EDH17B2; estradiol 17-beta-dehydrogenase 2; HSD17; HSD17B2; hydroxysteroid (17-beta) dehydrogenase 2; hydroxysteroid 17-beta dehydrogenase 2; microsomal 17-beta-hydroxysteroid dehydrogenase; SDR9C2; short chain dehydrogenase/reductase family 9C member 2; short chain dehydrogenase/reductase family 9C, member 2; tes; testosterone 17-beta-dehydrogenase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3294 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P37059 | |
| HSD17B2 | |
| Recombinant protein corresponding to Human HSD17B2. Recombinant protein control fragment (Product #RP-90922). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction