missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSD17B13 Recombinant Rabbit Monoclonal Antibody (10T10L5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551368
This item is not returnable.
View return policy
Description
Immunogen sequence: RRKSVAGEIV LITGAGHGIG RQTTYEFAKR QSILVLWDIN KRGVEETAAE CRKLGVTAHA YVVDCSNREE IYRSLNQVKK EVGDVTIVVN NAGTVYPADL LSTKDEEITK TFEVNILGHF WITKALLPSM MERNHGHIVT VASVCGHEGI PYLIPYCSSK FAAVGFHRGL TSELQALGKT GI.
HSD17B13 (Hydroxysteroid 17 Beta Dehydrogenase 13) is a Protein Coding gene. Diseases associated with HSD17B13 include5 Alpha Reductase DeficiencyandMucopolysaccharidosis Iii. Among its related pathways areMetabolismandLipoprotein metabolism. Gene Ontology (GO) annotations related to this gene includeoxidoreductase activity. An important paralog of this gene isHSD17B11.
Specifications
| HSD17B13 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q7Z5P4 | |
| HSD17B13 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
| ELISA, Western Blot | |
| 10T10L5 | |
| Unconjugated | |
| HSD17B13 | |
| 17-beta hydroxysteroid dehydrogenase; 17-beta hydroxysteroid dehydrogenase 13; 17-beta-HSD 13; 17-beta-hydroxysteroid dehydrogenase 13; AI047820; Alcohol dehydrogenase PAN1B-like; HMFN0376; Hsd17b13; hydroxysteroid (17-beta) dehydrogenase 13; hydroxysteroid 17-beta dehydrogenase 13; NIIL497; Pan1b; PAN1B-like; SCDR9; SDR16C3; short chain dehydrogenase/reductase family 16C member 3; short chain dehydrogenase/reductase family 16C, member 3; short-chain dehydrogenase/reductase 9; UNQ497/PRO1014 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 345275 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction