missing translation for 'onlineSavingsMsg'
Learn More

HSD17B13 Antibody (HSD17B13/13109), Novus Biologicals™

Mouse Monoclonal Antibody

£195.00 - £368.00

Specifications

Antigen HSD17B13
Clone HSD17B13/13109
Concentration 0.2 mg/ml
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30290095
View Documents
Novus Biologicals
NBP3-44611-20ug
20 μg
£195.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30301962
View Documents
Novus Biologicals
NBP3-44611-100ug
100 μg
£368.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HSD17B13 Monoclonal antibody specifically detects HSD17B13 in Human samples. It is validated for nan
Specifications

Specifications

HSD17B13
0.2 mg/ml
Monoclonal
Purified
RUO
Human
HMFN0376, hydroxysteroid (17-beta) dehydrogenase 13, member 3,17-beta-hydroxysteroid dehydrogenase 13, MGC138508,17-beta-HSD 13, NIIL497, SCDR9, short-chain dehydrogenase/reductase 9, UNQ497/PRO1014
Recombinant fragment (around aa1-200) of human HSD17B13 protein (Uniprot: Q7Z5P4). Exact sequence is proprietary.
Primary
Store at 4⪚C.
HSD17B13/13109
Immunohistochemistry
Unconjugated
Mouse
Cell Biology, Endocrinology, Signal Transduction
10 mM PBS with 0.05% BSA
345275
IgG2b κ
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.