missing translation for 'onlineSavingsMsg'
Learn More

HSD17B13 Antibody (HSD17B13/13101), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-44605-100ug

 View more versions of this product

Product Code. 30291605

  • £368.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HSD17B13 Monoclonal antibody specifically detects HSD17B13 in Human samples. It is validated for nan
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSD17B13
Monoclonal
0.2 mg/ml
10 mM PBS with 0.05% BSA
Mouse
Protein A purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry
HSD17B13/13101
Unconjugated
HMFN0376, hydroxysteroid (17-beta) dehydrogenase 13, member 3,17-beta-hydroxysteroid dehydrogenase 13, MGC138508,17-beta-HSD 13, NIIL497, SCDR9, short-chain dehydrogenase/reductase 9, UNQ497/PRO1014
Recombinant fragment (around aa1-200) of human HSD17B13 protein (Uniprot: Q7Z5P4). Exact sequence is proprietary.
100 μg
Cell Biology, Endocrinology, Signal Transduction
345275
Store at 4⪚C.
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.