missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSD17B12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553418
This item is not returnable.
View return policy
Description
Immunogen sequence: WGVGNEAGVG PGLGEWAVVT GSTDGIGKSY AEELAKHGMK VVLISRSKDK LDQVSSEIKE KFKVETRTIA VDFASEDIYD KIKTGLAGLE IGILVNNVGM SYEYPEYFLD VPDLD Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.
Specifications
| HSD17B12 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B12 | |
| 17-beta-HSD 12; 17beta-HSD type 12; 17-beta-hydroxysteroid dehydrogenase 12; 2610510O05Rik; 3-ketoacyl-CoA reductase; AI172963; Estradiol 17-beta-dehydrogenase 12; HSD17B12; hydroxysteroid (17-beta) dehydrogenase 12; hydroxysteroid 17-beta dehydrogenase 12; KAR; keratoadhesin; keratonectin; Kik1; KIK-I; SDR12C1; short chain dehydrogenase/reductase family 12C member 1; short chain dehydrogenase/reductase family 12C, member 1; smooth muscle-specific 17 beta-hydroxysteroid dehydrogenase type 3; steroid dehydrogenase; steroid dehydrogenase homolog; very-long-chain 3-oxoacyl-CoA reductase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51144 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53GQ0 | |
| HSD17B12 | |
| Recombinant protein corresponding to Human HSD17B12. Recombinant protein control fragment (Product #RP-91081). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction