Learn More
Invitrogen™ HSD17B12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595888
Description
Immunogen sequence: AGVGPGLGEW AVVTGSTDGI GKSYAEELAK HGMKVVLISR SKDKLDQVSS EIKEKFKVET RTIAVDFASE DIYDKIKTGL AGLEIGILVN NVGMSYEYPE YFLDVPDLDN VIKKMININI LSVCKMTQLV LPGMVERSKG AILNISSGSG MLPVPLLTIY SATKTFVDFF SQCLHEEYRS KGVFVQSVLP YFVATKLAKI RKPTLDKPSP ETFVKSAIKT VGLQSRTNGY LIHALMGSII SNLPSWIYLK IVMNMNKSTR AHYLKKTKKN.
The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.
Specifications
| HSD17B12 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B12 | |
| 17-beta-HSD 12; 17beta-HSD type 12; 17-beta-hydroxysteroid dehydrogenase 12; 2610510O05Rik; 3-ketoacyl-CoA reductase; AI172963; Estradiol 17-beta-dehydrogenase 12; HSD17B12; hydroxysteroid (17-beta) dehydrogenase 12; hydroxysteroid 17-beta dehydrogenase 12; KAR; keratoadhesin; keratonectin; Kik1; KIK-I; SDR12C1; short chain dehydrogenase/reductase family 12C member 1; short chain dehydrogenase/reductase family 12C, member 1; smooth muscle-specific 17 beta-hydroxysteroid dehydrogenase type 3; steroid dehydrogenase; steroid dehydrogenase homolog; very-long-chain 3-oxoacyl-CoA reductase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51144 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.29 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q53GQ0 | |
| HSD17B12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 43-312 of human HSD17B12 (NP_0572261). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.