missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17301-1-AP-150UL
This item is not returnable.
View return policy
Description
Short-chain alcohol dehydrogenases, such as HSD17B11, metabolize secondary alcohols and ketones.Specifications
| HSD17B11 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B11 | |
| 17 beta HSD 11, 17 beta HSD XI, 17 BETA HSD11, 17 BETA HSDXI, 17betaHSD11, 17betaHSDXI, 17bHSD11, DHRS8, HSD17B11, PAN1B, RETSDR2, SDR16C2, UNQ207/PRO233 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114664, 289456, 51170 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6AYS8, Q8NBQ5, Q9EQ06 | |
| Hsd17b11 | |
| HSD17B11 Fusion Protein Ag11182 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction