Learn More
HSD17B11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16303-1-AP-150UL
 View more versions of this product
Product Code. 16875894
  • £349.00 / 150µL
Description
Short-chain alcohol dehydrogenases, such as HSD17B11, metabolize secondary alcohols and ketones.
Specifications
Specifications
HSD17B11
Polyclonal
Unconjugated
HSD17B11
17 beta HSD 11, 17 beta HSD XI, 17 BETA HSD11, 17 BETA HSDXI, 17betaHSD11, 17betaHSDXI, 17bHSD11, DHRS8, HSD17B11, PAN1B, RETSDR2, SDR16C2, UNQ207/PRO233
Rabbit
Antigen Affinity Chromatography
RUO
114664, 289456, 51170
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6AYS8, Q8NBQ5, Q9EQ06
Hsd17b11
HSD17B11 Fusion Protein Ag9278
150 μL
Primary
Human, Mouse, Rat