missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSCB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553802
This item is not returnable.
View return policy
Description
Immunogen sequence: QFLIEIMEIN EKLAEAESEA AMKEIESIVK AKQKEFTDNV SSAFEQDDFE EAKEILTKMR YFSNIEEKIK LKK Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 79%.
May act as a co-chaperone in iron-sulfur cluster assembly in mitochondria.
Specifications
| HSCB | |
| Polyclonal | |
| Unconjugated | |
| HSCB | |
| AI325508; AW049829; C-HSC20; dJ366L4.2; DnaJ (Hsp40) homolog, subfamily C, member 20; DnaJ homolog subfamily C member 20; DNAJC20; HSC20; HSCB; HscB iron-sulfur cluster co-chaperone homolog; HscB iron-sulfur cluster co-chaperone homolog (E. coli); HscB mitochondrial iron-sulfur cluster cochaperone; HscB mitochondrial iron-sulfur cluster co-chaperone; iron-sulfur cluster co-chaperone protein HscB; Iron-sulfur cluster co-chaperone protein HscB, cytoplasmic; Iron-sulfur cluster co-chaperone protein HscB, mitochondrial; JAC1; J-type co-chaperone HSC20; RGD1311005 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 150274 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IWL3 | |
| HSCB | |
| Recombinant protein corresponding to Human HSCB. Recombinant protein control fragment (Product #RP-92350). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction