missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSCB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15132-1-AP-20UL
This item is not returnable.
View return policy
Description
May act as a co-chaperone in iron-sulfur cluster assembly in mitochondria.Specifications
| HSCB | |
| Polyclonal | |
| Unconjugated | |
| HSCB | |
| DNAJC20, HSC20, HSCB, JAC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 150274 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8IWL3 | |
| HSCB | |
| HSCB Fusion Protein Ag7268 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction