missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSCB Antibody (OTI3E1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72415PE
This item is not returnable.
View return policy
Description
HSCB Monoclonal antibody specifically detects HSCB in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| HSCB | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI3E1 | |
| Western Blot, Immunohistochemistry | |
| DnaJ homolog subfamily C member 20, DNAJC20DnaJ (Hsp40) homolog, subfamily C, member 20, Hsc20, HSC20dJ366L4.2, HscB iron-sulfur cluster co-chaperone homolog (E. coli), iron-sulfur cluster co-chaperone protein HscB, mitochondrial, Jac1, J-type co-chaperone HSC20, MGC2637, MGC74462 | |
| Full length human recombinant protein of human HSCB(NP_741999) produced in HEK293T cell | |
| 0.1 mL | |
| metabolism | |
| 150274 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction