missing translation for 'onlineSavingsMsg'
Learn More

HSCB Antibody (OTI3E1), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18421558 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421558 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421558 Supplier Novus Biologicals Supplier No. NBP272415AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HSCB Monoclonal antibody specifically detects HSCB in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSCB
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI3E1
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias DnaJ homolog subfamily C member 20, DNAJC20DnaJ (Hsp40) homolog, subfamily C, member 20, Hsc20, HSC20dJ366L4.2, HscB iron-sulfur cluster co-chaperone homolog (E. coli), iron-sulfur cluster co-chaperone protein HscB, mitochondrial, Jac1, J-type co-chaperone HSC20, MGC2637, MGC74462
Host Species Mouse
Immunogen Full length recombinant protein of human HSCB (NP_741999) produced in HEK293T cells
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 150274
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.