missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsc70 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81719-2-RR-100UL
This item is not returnable.
View return policy
Description
The Hsc70 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Hsc70. This antibody recognizes human, mouse, rat antigen. The Hsc70 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| Hsc70 | |
| Recombinant Monoclonal | |
| 500 μg/mL | |
| HSC54; HSC70; HSC71; HSP71; HSP73; HSPA10; LAP1; MGC131511; MGC29929; NIP71 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 70 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 3H9 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3312 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction