missing translation for 'onlineSavingsMsg'
Learn More

Hsc70 Rabbit Recombinant Antibody, Proteintech

Rabbit Recombinant Antibody

Brand:  Proteintech 81719-2-RR-20UL

 View more versions of this product

Product Code. 30235652

  • ONLINE EXCLUSIVE PRICE
    £135.00
    £119.00 / 20µL
    Save £16.00
    11% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Hsc70 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Hsc70. This antibody recognizes human, mouse, rat antigen. The Hsc70 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications

Specifications

Hsc70
Recombinant Monoclonal
500 μg/mL
HSC54; HSC70; HSC71; HSP71; HSP73; HSPA10; LAP1; MGC131511; MGC29929; NIP71
Recombinant Protein
20 μL
Primary
Human, Mouse, Rat
70 kDa
Liquid
Western Blot, ELISA
3H9
Unconjugated
Rabbit
Protein A purification
RUO
3312
Store at -20°C. Stable for one year after shipment.
Recombinant Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.