missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HS747E2A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-17262-100UL
This item is not returnable.
View return policy
Description
HS747E2A Polyclonal antibody specifically detects HS747E2A in Human samples. It is validated for Immunofluorescence
Specifications
| HS747E2A | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
| bK747E2.1, chromosome 22 open reading frame 31, HS747E2A, hypothetical protein LOC25770 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: HPINVRRDPSIPIYGLRQSILLNTRLQDCYVDSPALTNIWMARTCAKQNINAPAPATTSSWEVVRNPLIASSFSLVKLVLRRQLKNKCCPPPCKFGEGKLSKRLKHKDDSVMKATQQARKRNFISSKSKQPAGHRR | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 25770 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction