Learn More
HS6ST2 Polyclonal Antibody
GREENER_CHOICE
HS6ST2 Polyclonal Antibody
Brand:  Invitrogen PA547372
Product Code. 15567862
  • £442.00 / 100µg
Description
HS6ST2 Polyclonal Antibody for Western Blot, IP
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HS6ST2
Polyclonal
Unconjugated
HS6ST2
6OST2; EC-1; heparan sulfate 6-O-sulfotransferase 2; heparan-sulfate 6-O-sulfotransferase 2; Hs6st2; HS6ST-2; LOW QUALITY PROTEIN: heparan-sulfate 6-O-sulfotransferase 2; PSEC0092; RGD1563872; RGD1564397
Goat
Antigen affinity chromatography
RUO
90161
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
Q96MM7
HS6ST2
Mouse myeloma cell line NS0-derived recombinant human Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 Gln179-Arg605.