missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HS3ST5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584662
This item is not returnable.
View return policy
Description
Immunogen sequence: DYTQVLEGKE RKNKTYYKFE KLAIDPNTCE VNTKYKAVRT SIYTKHLERW LKYFPIEQFH VVDGDRLITE PLP Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.
HS3ST5 belongs to a group of heparan sulfate 3-O-sulfotransferases (EC 2.8.2.23) that transfer sulfate from 3-prime-phosphoadenosine 5-prime phosphosulfate (PAPS) to heparan sulfate and heparin (Mochizuki et al., 2003 [PubMed 12740361]).[supplied by OMIM, Mar 2008].
Specifications
| HS3ST5 | |
| Polyclonal | |
| Unconjugated | |
| HS3ST5 | |
| 3OST5; 3-OST-5; D930005L05Rik; Gm1151; h3-OST-5; heparan sulfate (glucosamine) 3-O-sulfotransferase 5; heparan sulfate (glucosamine) 3-O-sulfotransferase 6; heparan sulfate 3-OST-5; heparan sulfate 3-O-sulfotransferase 5; heparan sulfate D-glucosaminyl 3-O-sulfotransferase 5; Heparan sulfate glucosamine 3-O-sulfotransferase 5; heparan sulfate-glucosamine 3-sulfotransferase 5; Hs3ost5; HS3ST5; NBLA04021 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 222537 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZT8 | |
| HS3ST5 | |
| Recombinant protein corresponding to Human HS3ST5. Recombinant protein control fragment (Product #RP-105330). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction