missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HS3ST4 Monoclonal Antibody (712010)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524332
This item is not returnable.
View return policy
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
| HS3ST4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9Y661 | |
| HS3ST4 | |
| CHO-derived recombinant human Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Gly184-Lys456. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 712010 | |
| Unconjugated | |
| HS3ST4 | |
| 30ST4; 3OST4; 3-OST-4; EC 2.8.2; EC 2.8.2.23; EG628779; Gm6915; h3-OST-4; heparan sulfate (glucosamine) 3-O-sulfotransferase 4; heparan sulfate 3-O-sulfotransferase 4; heparan sulfate 3-O-sulfotransferase-4; heparan sulfate D-glucosaminyl 3-O-sulfotransferase 4; Heparan sulfate glucosamine 3-O-sulfotransferase 4; heparan sulfate-glucosamine 3-sulfotransferase 4; Hs3st4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 9951 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction