missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HS3ST3B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566664
This item is not returnable.
View return policy
Description
Immunogen sequence: LLLGSGSRAAH DPPALATAPD GTPPRLPFRA PPATPLASGK EMAEGAASPE EQSPEVPDSP SPISSFFSGS GSK Highest antigen sequence identity to the following orthologs - mouse 73%, rat 72%.
HS3ST3B1 is a member of the heparan sulfate biosynthetic enzyme family. It is a type II integral membrane protein and possesses heparan sulfate glucosaminyl 3-O-sulfotransferase activity. The sulfotransferase domain of this enzyme is highly similar to the same domain of heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3A1, and these two enzymes sulfate an identical disaccharide. This gene is widely expressed, with the most abundant expression in liver and placenta.Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. The enzyme encoded by this gene is a member of the heparan sulfate biosynthetic enzyme family. It is a type II integral membrane protein and possesses heparan sulfate glucosaminyl 3-O-sulfotransferase activity. The sulfotransferase domain of this enzyme is highly similar to the same domain of heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3A1, and these two enzymes sulfate an identical disaccharide. This gene is widely expressed, with the most abundant expression in liver and placenta.
Specifications
| HS3ST3B1 | |
| Polyclonal | |
| Unconjugated | |
| HS3ST3B1 | |
| 30ST3B1; 3-OST-3B; 3OST3B1; h3-OST-3B; heparan sulfate (glucosamine) 3-O-sulfotransferase 3B1; heparan sulfate 3-O-sulfotransferase 3B1; heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3B1; Heparan sulfate glucosamine 3-O-sulfotransferase 3B1; heparan sulfate-glucosamine 3-sulfotransferase 3B1; HS3ST3B; HS3ST3B1; LOC100724084 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9953 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y662 | |
| HS3ST3B1 | |
| Recombinant Human HS3ST3B1. Recombinant protein control fragment (Product #RP-105300). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction